4-Bromo-2-iodophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4957-1G
B4957-5G
TCB4957-1GEA
169.06
CAD
TCB4957-1G
TCB4957-5G
4-Bromo-2-iodophenol ≥98.0% (by GC, titration analysis)
4-Bromo-2-iodophenol
Formula:
C₆H₄BrIO MW: 298.91 g/mol |
MDL Number:
MFCD09029644 CAS Number: 207115-22-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...