Diethyl (4-Bromophenyl)phosphonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (4-Bromophenyl)phosphonic acid Diethyl ester

D5453-1G D5453-200MG
TCD5453-1GEA 455.04 CAD
TCD5453-1G TCD5453-200MG
Diethyl (4-Bromophenyl)phosphonate ≥98.0% (by GC)
Diethyl (4-Bromophenyl)phosphonate
Formula: C₁₀H₁₄BrO₃P
MW: 293.10 g/mol
Boiling Pt: 120°C
MDL Number: MFCD00769296
CAS Number: 20677-12-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR