2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

T3318-1G T3318-5G
76220-558EA 101.12 CAD
76220-558 76220-560
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl
Formula: C₂₁H₁₆F₄
MDL Number: MFCD12964272
CAS Number: 205806-87-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 61.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR