Oxibendazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: (5-Propoxy-1H-benzimidazol-2-yl)carbamic acid methyl ester, Methyl (5-Propoxy-1H-benzo[d]imidazol-2-yl)carbamate

O0499-100MG O0499-500MG
TCO0499-100MGEA 97.96 CAD
TCO0499-100MG TCO0499-500MG
Oxibendazole ≥98.0% (by HPLC, total nitrogen)
Oxibendazole
Formula: C₁₂H₁₅N₃O₃
MW: 249.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133728
CAS Number: 20559-55-1
EINECS: 243-877-7
Merck Index: 14,06936

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR