Benzo[b]naphtho[1,2-d]thiophene ≥98.0% (by GC)

Supplier: TCI America

B4951-1G B4951-5G
TCB4951-1GEA 249.64 CAD
TCB4951-1G TCB4951-5G
Benzo[b]naphtho[1,2-d]thiophene ≥98.0% (by GC)
Benzo[b]naphtho[1,2-d]thiophene
Formula: C₁₆H₁₀S
Boiling Pt: 220°C
Storage Temperature: Refrigerator
MDL Number: MFCD00215942
CAS Number: 205-43-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR