9,9-Diphenyl-9,10-dihydroacridine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 9,10-Dihydro-9,9-diphenylacridine

D5332-1G D5332-200MG
TCD5332-1GEA 410.8 CAD
TCD5332-1G TCD5332-200MG
9,9-Diphenyl-9,10-dihydroacridine ≥98.0% (by HPLC, total nitrogen)
9,9-Diphenyl-9,10-dihydroacridine
Formula: C₂₅H₁₉N
MW: 333.43 g/mol
MDL Number: MFCD28127357
CAS Number: 20474-15-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 249.0 to 254.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR