Tributyl(ethyl)phosphonium Diethyl Phosphate ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Tributyl(ethyl)phosphonium diethylphosphate

T3641-25G T3641-5G
TCT3641-25GEA 431.34 CAD
TCT3641-25G TCT3641-5G
Tributyl(ethyl)phosphonium Diethyl Phosphate ≥96.0% (by titrimetric analysis)
Tributyl(ethyl)phosphonium Diethyl Phosphate
Formula: C₁₈H₄₂O₄P₂
MW: 384.48 g/mol
MDL Number: MFCD17018975
CAS Number: 20445-94-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR