Cesium Pivalate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3230-1G C3230-5G
TCC3230-1GEA 82.16 CAD
TCC3230-1G TCC3230-5G
Cesium Pivalate ≥97.0% (by titrimetric analysis)
Cesium Pivalate
Formula: C₅H₉CsO₂
Melting Pt: 348°C
MDL Number: MFCD10567040
CAS Number: 20442-70-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR