N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N,N'-Bis(4-methylphenyl)-N,N'-diphenylbenzidine, 4,4'-Bis[N-phenyl-N-(p-tolyl)amino]biphenyl

D4834-1G D4834-200MG
TCD4834-1GEA 543.84 CAD
TCD4834-1G TCD4834-200MG
N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine ≥98.0% (by GC)
N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine
Formula: C₃₈H₃₂N₂
MW: 516.69 g/mol
MDL Number: MFCD00799301
CAS Number: 20441-06-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR