Formaldehyde Dimethylhydrazone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

F0713-1G F0713-5G
TCF0713-1GEA 189.6 CAD
TCF0713-1G TCF0713-5G
Formaldehyde Dimethylhydrazone ≥98.0% (by GC, titration analysis)
Formaldehyde Dimethylhydrazone
Formula: C₃H₈N₂
MW: 72.11 g/mol
Boiling Pt: 71°C
Melting Pt: -103°C
Storage Temperature: Refrigerator
MDL Number: MFCD01753011
CAS Number: 2035-89-4
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR