2,4-Dibromophenyl Glycidyl ether ≥90.0% (by GC)

Supplier: TCI America
Warning

D4885-25G D4885-500G
TCD4885-25GEA 60.04 CAD
TCD4885-25G TCD4885-500G
2,4-Dibromophenyl Glycidyl ether ≥90.0% (by GC)
2,4-Dibromophenyl Glycidyl ether
Formula: C₉H₈Br₂O₂
MDL Number: MFCD13194826
CAS Number: 20217-01-0
EINECS: 243-606-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR