Diisopropyl Hydrazine-1,2-dicarboxylate ≥98.0% (by GC)

Supplier: TCI America

D5365-1G D5365-200MG
TCD5365-1GEA 527.72 CAD
TCD5365-1G TCD5365-200MG
Diisopropyl Hydrazine-1,2-dicarboxylate ≥98.0% (by GC)
Diisopropyl Hydrazine-1,2-dicarboxylate
Formula: C₈H₁₆N₂O₄
Melting Pt: 108°C
MDL Number: MFCD19443326
CAS Number: 19740-72-8
EINECS: 243-263-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR