4-Methylumbelliferyl beta-D-ribofuranoside ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
7-(((2S,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-4-methyl-2H-chromen-2-one, 4-Methylumbelliferyl β-D-Ribofuranoside
M3027-100MG
M3027-25MG
TCM3027-100MGEA
788.42
CAD
TCM3027-100MG
TCM3027-25MG
4-Methylumbelliferyl beta-D-ribofuranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl beta-D-ribofuranoside
Formula:
C₁₅H₁₆O₇ MW: 308.29 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD06796941 CAS Number: 195385-93-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -147 to -152 deg (C=0.15, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...