2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4938-1G B4938-5G
TCB4938-1GEA 357.96 CAD
TCB4938-1G TCB4938-5G
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran ≥98.0% (by HPLC, titration analysis)
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran
Formula: C₁₉H₁₆I₂O₃
MDL Number: MFCD02675787
CAS Number: 1951-26-4
EINECS: 217-773-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR