2-Iodo-2-methylpropionitrile ≥96.0% (by GC)

Supplier: TCI America
Danger

I0954-1G I0954-5G
TCI0954-1GEA 171.9 CAD
TCI0954-1G TCI0954-5G
2-Iodo-2-methylpropionitrile ≥96.0% (by GC)
2-Iodo-2-methylpropionitrile
Formula: C₄H₆IN
Boiling Pt: 62°C
Storage Temperature: Freezer
CAS Number: 19481-79-9
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR