4-Bromo-N-methoxy-N-methylbenzamide ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
B5728-1G
B5728-5G
TCB5728-1GEA
102.7
CAD
TCB5728-1G
TCB5728-5G
4-Bromo-N-methoxy-N-methylbenzamide ≥95.0% (by GC)
4-Bromo-N-methoxy-N-methylbenzamide
Formula:
C₉H₁₀BrNO₂ MW: 244.09 g/mol |
MDL Number:
MFCD02684303 CAS Number: 192436-83-2 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...