Pentafluorophenyl Acetate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
P2192-1G
P2192-5G
TCP2192-1GEA
150.1
CAD
TCP2192-1G
TCP2192-5G
Pentafluorophenyl Acetate ≥98.0% (by GC)
Pentafluorophenyl Acetate
Formula:
C₈H₃F₅O₂ Boiling Pt: 60°C |
MDL Number:
MFCD00015552 CAS Number: 19220-93-0 EINECS: 242-891-0 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 30.0 to 34.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...