1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T3537-1G T3537-200MG
76220-668EA 521.4 CAD
76220-668 76220-670
1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene ≥98.0% (by HPLC, titration analysis)
1,3,5-Tris(1-phenyl-1H-benzimidazol-2-yl)benzene
Formula: C₄₅H₃₀N₆
MDL Number: MFCD08276433
CAS Number: 192198-85-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR