9,10-Bis[N-(p-tolyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5149-1G B5149-200MG
TCB5149-1GEA 677.82 CAD
TCB5149-1G TCB5149-200MG
9,10-Bis[N-(p-tolyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)
9,10-Bis[N-(p-tolyl)anilino]anthracene
Formula: C₄₀H₃₂N₂
MDL Number: MFCD12022466
CAS Number: 190974-21-1

Order Now

Specification Test Results

Appearance Yellow to Amber powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR