1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane ≥95.0% (by GC)
Supplier: TCI America
T3548-1ML
T3548-5ML
76220-684EA
80.58
CAD
76220-684
76220-688
1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane ≥95.0% (by GC)
1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane
Formula:
C₁₄H₄₄O₆Si₇ Boiling Pt: 128°C |
CAS Number:
19095-23-9 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...