4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98,0 GC_ASSAY_METHOD

Supplier: TCI America

Synonyms: 4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

E1158-25G E1158-5G
TCE1158-25GEA 744.18 CAD
TCE1158-25G TCE1158-5G
4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98,0 GC_ASSAY_METHOD
4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl
Formula: C₂₃H₂₈F₂O
MW: 358.47 g/mol
MDL Number: MFCD18451991
CAS Number: 189750-98-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C
Mesomorphic range 77.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR