9,10-Bis[N-(m-tolyl)anilino]anthracene ≥98.0% (by HPLC)

Supplier: TCI America

B4965-1G B4965-200MG
TCB4965-1GEA 398.32 CAD
TCB4965-1G TCB4965-200MG
9,10-Bis[N-(m-tolyl)anilino]anthracene ≥98.0% (by HPLC)
9,10-Bis[N-(m-tolyl)anilino]anthracene
Formula: C₄₀H₃₂N₂
MDL Number: MFCD12022466
CAS Number: 189263-81-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR