1-Butylpyridinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B5571-25G
B5571-5G
76218-848EA
521.4
CAD
76218-848
76218-850
1-Butylpyridinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Butylpyridinium bis(trifluoromethanesulfonyl)imide
Formula:
C₁₁H₁₄F₆N₂O₄S₂ |
MDL Number:
MFCD16038143 CAS Number: 187863-42-9 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...