3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D5421-1G D5421-200MG
TCD5421-1GEA 421.86 CAD
TCD5421-1G TCD5421-200MG
3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole
Formula: C₂₀H₁₅Br₂N
MDL Number: MFCD31618134
CAS Number: 1873364-08-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 239.0 to 243.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR