[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥97.0% (by HPLC)
Supplier: TCI America
|
Warning
|
F1111-1G
F1111-200MG
76220-096EA
467.68
CAD
76220-096
76220-098
[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥97.0% (by HPLC)
[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
Formula:
C₂₃H₂₁F₄IO₆S Storage Temperature: Refrigerator |
MDL Number:
MFCD30475655 CAS Number: 1868173-29-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 % (total of Ion-Pair) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...