tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate ≥97.0% (by GC)

Supplier: TCI America

B5587-1G B5587-5G
TCB5587-1GEA 109.02 CAD
TCB5587-1G TCB5587-5G
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate ≥97.0% (by GC)
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate
Formula: C₁₃H₂₆O₆
MDL Number: MFCD03791106
CAS Number: 186020-66-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Acid Value max. 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR