(3-Bromophenyl)triphenylsilane ≥98.0% (by GC)

Supplier: TCI America

B5222-1G B5222-200MG
76219-530EA 314.42 CAD
76219-530 76219-532
(3-Bromophenyl)triphenylsilane ≥98.0% (by GC)
(3-Bromophenyl)triphenylsilane
Formula: C₂₄H₁₉BrSi
MDL Number: MFCD28098175
CAS Number: 185626-73-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR