1-(2-Propynyloxy)naphthalene ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Propargyloxy)naphthalene

P2227-1G P2227-5G
TCP2227-1GEA 680.98 CAD
TCP2227-1G TCP2227-5G
1-(2-Propynyloxy)naphthalene ≥95.0% (by GC)
1-(2-Propynyloxy)naphthalene
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 75°C
Storage Temperature: Refrigerator
MDL Number: MFCD03787730
CAS Number: 18542-45-5

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR