Methyl 4-Bromo-2-chlorobenzoate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2883-1G
M2883-5G
TCM2883-1GEA
72.68
CAD
TCM2883-1G
TCM2883-5G
Methyl 4-Bromo-2-chlorobenzoate ≥98.0% (by GC)
Methyl 4-Bromo-2-chlorobenzoate
Formula:
C₈H₆BrClO₂ |
MDL Number:
MFCD10566823 CAS Number: 185312-82-7 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...