Ethyl 3,4-Bis(2-methoxyethoxy)benzoate ≥98.0% (by GC)

Supplier: TCI America

E1186-1G E1186-5G
TCE1186-1GEA 53.72 CAD
TCE1186-1G TCE1186-5G
Ethyl 3,4-Bis(2-methoxyethoxy)benzoate ≥98.0% (by GC)
Ethyl 3,4-Bis(2-methoxyethoxy)benzoate
Formula: C₁₅H₂₂O₆
MDL Number: MFCD01935284
CAS Number: 183322-16-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR