(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3174-1G T3174-5G
TCT3174-1GEA 89.27 CAD
TCT3174-1G TCT3174-5G
(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) ≥98.0% (by HPLC)
(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers)
Formula: C₈H₄F₃N₃O
MDL Number: MFCD00593044
CAS Number: 183266-61-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR