Docosyl acrylate ≥95.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D5075-100G D5075-25G
TCD5075-100GEA 205.4 CAD
TCD5075-100G TCD5075-25G
Docosyl acrylate ≥95.0% (by GC) stabilized
Docosyl acrylate
Formula: C₂₅H₄₈O₂
MDL Number: MFCD00080409
CAS Number: 18299-85-9
EINECS: 242-182-6

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR