1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: TMS-EBX

T3272-1G T3272-5G
76220-544EA 322.32 CAD
76220-544 76220-546
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0% (by titrimetric analysis)
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one
Formula: C₁₂H₁₃IO₂Si
MW: 344.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29765750
CAS Number: 181934-29-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Potassium iodate Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR