(3-Chloropropyl)tris(trimethylsilyloxy)silane ≥96.0% (by GC)

Supplier: TCI America
Warning

C3437-1G C3437-5G
76218-940EA 44.24 CAD
76218-940 76218-942
(3-Chloropropyl)tris(trimethylsilyloxy)silane ≥96.0% (by GC)
(3-Chloropropyl)tris(trimethylsilyloxy)silane
Formula: C₁₂H₃₃ClO₃Si₄
Boiling Pt: 181°C
Flash Pt: 85°C
MDL Number: MFCD00054781
CAS Number: 18077-31-1
EINECS: 241-985-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR