2,5-Dibromo-3-hexadecylthiophene ≥96.0% (by GC)

Supplier: TCI America

D5242-1G D5242-5G
76219-866EA 180.12 CAD
76219-866 76219-868
2,5-Dibromo-3-hexadecylthiophene ≥96.0% (by GC)
2,5-Dibromo-3-hexadecylthiophene
Formula: C₂₀H₃₄Br₂S
Storage Temperature: Refrigerator
CAS Number: 178452-13-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 34.0 to 38.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR