3-Amino-4-bromo-5-methylpyrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Bromo-5-methyl-1H-pyrazol-3-amine

A2970-1G A2970-5G
TCA2970-1GEA 112.18 CAD
TCA2970-1G TCA2970-5G
3-Amino-4-bromo-5-methylpyrazole ≥98.0% (by HPLC, titration analysis)
3-Amino-4-bromo-5-methylpyrazole
Formula: C₄H₆BrN₃
MW: 176.02 g/mol
MDL Number: MFCD03412169
CAS Number: 1780-72-9

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR