3-Chloro-2-ethoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3081-1G C3081-5G
TCC3081-1GEA 279.66 CAD
TCC3081-1G TCC3081-5G
3-Chloro-2-ethoxypyridine ≥98.0% (by GC, titration analysis)
3-Chloro-2-ethoxypyridine
Formula: C₇H₈ClNO
MDL Number: MFCD09702462
CAS Number: 177743-06-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR