4-Biphenylyl trifluoromethanesulfonate ≥98.0% (by GC)

Supplier: TCI America
Warning

B5012-1G B5012-5G
TCB5012-1GEA 75.84 CAD
TCB5012-1G TCB5012-5G
4-Biphenylyl trifluoromethanesulfonate ≥98.0% (by GC)
4-Biphenylyl trifluoromethanesulfonate
Formula: C₁₃H₉F₃O₃S
MDL Number: MFCD18379753
CAS Number: 17763-78-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR