5'-Bromo-[2,2'-bithiophene]-5-carbonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5457-1G
B5457-200MG
TCB5457-1GEA
478.74
CAD
TCB5457-1G
TCB5457-200MG
5'-Bromo-[2,2'-bithiophene]-5-carbonitrile ≥98.0% (by GC)
5'-Bromo-[2,2'-bithiophene]-5-carbonitrile
Formula:
C₉H₄BrNS₂ |
CAS Number:
176787-96-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...