trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

D5127-1G D5127-5G
TCD5127-1GEA 80.58 CAD
TCD5127-1G TCD5127-5G
trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl
Formula: C₂₁H₃₀F₂
MDL Number: MFCD26143186
CAS Number: 174350-08-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 55.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR