2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
H1546-1G
H1546-5G
76220-132EA
154.84
CAD
76220-132
76220-134
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione ≥98.0% (by GC)
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione
Formula:
C₁₃H₂₀O₃ |
MDL Number:
MFCD01862890 CAS Number: 172611-72-2 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...