9,9',10,10'-Tetraphenyl-2,2'-bianthracene ≥97.0% (by HPLC)

Supplier: TCI America

T3336-1G T3336-200MG
TCT3336-1GEA 529.3 CAD
TCT3336-1G TCT3336-200MG
9,9',10,10'-Tetraphenyl-2,2'-bianthracene ≥97.0% (by HPLC)
9,9',10,10'-Tetraphenyl-2,2'-bianthracene
Formula: C₅₂H₃₄
MW: 658.84 g/mol
CAS Number: 172285-72-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR