Lithium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

L0281-25G L0281-5G
TCL0281-25GEA 589.37 CAD
TCL0281-25G TCL0281-5G
Lithium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)
Lithium bis(fluorosulfonyl)imide
Formula: F₂LiNO₄S₂
MDL Number: MFCD28126786
CAS Number: 171611-11-3
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR