4-Bromo-2-methylbenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(4-Bromo-2-methylphenyl)methanol
B5231-1G
B5231-5G
TCB5231-1GEA
72.68
CAD
TCB5231-1G
TCB5231-5G
4-Bromo-2-methylbenzyl alcohol ≥98.0% (by GC)
4-Bromo-2-methylbenzyl alcohol
Formula:
C₈H₉BrO |
MDL Number:
MFCD11847398 CAS Number: 17100-58-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...