3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane ≥96.0% (by GC)

Supplier: TCI America
Warning

D5263-25ML D5263-5ML
TCD5263-25MLEA 238.58 CAD
TCD5263-25ML TCD5263-5ML
3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane ≥96.0% (by GC)
3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane
Formula: C₇H₂₄O₃Si₄
Boiling Pt: 164°C
MDL Number: MFCD00054789
CAS Number: 17082-46-1
EINECS: 241-136-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR