5-Aminovaleric acid Hydroiodide, low water content ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Aminopentanoic acid Hydroiodide, 5-AVAI, Homopiperidinic acid Hydroiodide, 5-Aminovaleric Acid Hydroiodide, 5-AVAI (Low water content), Homopiperidinic acid Hydroiodide (Low water content), 5-Aminopentanoic acid Hydroiodide (Low water content)
A2984-1G
A2984-5G
TCA2984-1GEA
222.78
CAD
TCA2984-1G
TCA2984-5G
5-Aminovaleric acid Hydroiodide, low water content ≥97.0% (by total nitrogen and titration analysis)
5-Aminovaleric acid Hydroiodide
Formula:
C₅H₁₁NO₂·HI MW: 245.06 g/mol |
CAS Number:
1705581-28-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Water | max. 200 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...