(4-Bromophenyl)phosphonic acid ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B5015-1G B5015-200MG
TCB5015-1GEA 1543.66 CAD
TCB5015-1G TCB5015-200MG
(4-Bromophenyl)phosphonic acid ≥98.0% (by HPLC)
(4-Bromophenyl)phosphonic acid
Formula: C₆H₆BrO₃P
MDL Number: MFCD00159409
CAS Number: 16839-13-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR