N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-phenyl-9H-fluoren-2-amine

B5045-1G B5045-200MG
TCB5045-1GEA 350.76 CAD
TCB5045-1G TCB5045-200MG
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline ≥98.0% (by total nitrogen basis)
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline
Formula: C₃₆H₃₁N
MW: 477.65 g/mol
MDL Number: MFCD29089346
CAS Number: 165320-27-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR