1,4-Dibromo-2,3,5,6-tetramethylbenzene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5168-1G
D5168-5G
76219-210EA
121.66
CAD
76219-210
76219-212
1,4-Dibromo-2,3,5,6-tetramethylbenzene ≥98.0% (by GC)
1,4-Dibromo-2,3,5,6-tetramethylbenzene
Formula:
C₁₀H₁₂Br₂ Boiling Pt: 317°C |
MDL Number:
MFCD00017787 CAS Number: 1646-54-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...