5-Azoniaspiro[4.4]nonane bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A3091-1G A3091-5G
TCA3091-1GEA 121.66 CAD
TCA3091-1G TCA3091-5G
5-Azoniaspiro[4.4]nonane bromide ≥98.0% (by titrimetric analysis)
5-Azoniaspiro[4.4]nonane bromide
Formula: C₈H₁₆BrN
Melting Pt: 273°C
MDL Number: MFCD30749246
CAS Number: 16450-38-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR